Skip to Content
MilliporeSigma

PMC3017

2-Hydroxypyridine N-oxide

≥98%, ISOM8™

Synonym(s):

2-Pyridinol 1-oxide, HOPO, Oxypyrion, 2-Hydroxypyridine N-oxide, 1-hydroxy-2-pyridone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C5H5NO2
CAS Number:
Molecular Weight:
111.10
UNSPSC Code:
12352005
EC Index Number:
235-100-8
NACRES:
NA.22
Beilstein/REAXYS Number:
1422545
MDL number:
PubChem Substance ID:
Assay:
≥98%
Form:
crystals, powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

reagent grade

Quality Segment

vapor pressure

~1.45X10-6 mmHg

product line

ISOM8™

assay

≥98%

form

crystals, powder

purified by

(HPLC)

reaction suitability

reaction type: Amidations, reaction type: C-N bond formation, reaction type: Complexometric reactions, reaction type: Coupling Reactions, reaction type: Peptide Synthesis

bp

387 °C

mp

147-152 °C

transition temp

flash point ~188.3 °C (~371 °F)

density

1.241 g/cm3 at 25 °C

application(s)

diagnostic assay manufacturing
life science and biopharma
pharmaceutical
pharmaceutical small molecule

storage temp.

15-25°C

SMILES string

O=N=1C=CC=CC1O

InChI

1S/C5H5NO2/c7-5-3-1-2-4-6(5)8/h1-4,7H

InChI key

JVHZMYAXZUIZKS-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8™ team, we have expertise and documentation to support ISO 9001, non-GMP, industrial needs, as defined by our M-Clarity™ Program. Our portfolio of ISOM8™ products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevate Program.

Application

Peptide Coupling and Synthesis:
  • Performance: 2-Hydroxypyridine N-oxide (HOPO) is recognized as a superior, non-explosive alternative to 1-hydroxybenzotriazole (HOBt) in carbodiimide-mediated amide bond formation.
  • Efficiency: Suppresses racemization during coupling reactions.
  • Solubility: It is generally soluble in water and organic solvents, making itversatile for various chemical reactions and applications.
  • Reactivity: Both functional groups hydroxyland N-oxide contributes to its reactivity, allowing it to participate invarious chemical transformations, including oxidation and coordination withmetal ions.
  • Stability: HOPO is generally stable under ambient conditions, although it canundergo tautomerism or other transformations depending on the reactionenvironment.
Metal Chelation and EnvironmentalApplications:
  • Ligand Properties: It can act as a bidentate ligand, coordinating to metal ionsthrough both the nitrogen and oxygen atoms.
  • Iron Sequestration: Effective in chelating iron, particularly from human transferrin.
  • Lanthanide Removal: Used for selective removal of lanthanides from environmental samples.
  • Electrochemical Sensors: HOPO has been used to modify gold electrodes for the sensitive detection of Europium, enabling effective monitoring in various applications.
Other Applications:
  • Coordination compound formation: 2-Hydroxypyridine N-oxide is used in coordination chemistry to form complexes with various transition metals. These metal complexes can have applications in catalysis, materials science, and as precursors for other chemical compounds.
  • Catalysis: It has been utilized as a ligand in catalytic systems,particularly in reactions involving transition metals, to enhance reaction rates and selectivity.
  • Pharmaceuticals: The compound has potential applications in medicinal chemistry as a building block or intermediate in the synthesis of biologically active compounds. Form stable complexes with metals can be leveraged in drug design and development.
Recent Studies and Innovations
  • Additive to supress racemization: HOPO is a critical coupling additivethat enhances the synthesis of active pharmaceutical ingredients (APIs) by suppressing racemization. Notably, it has been instrumental in developing nirmatrelvir, a potent inhibitor of the SARS-CoV-2 main protease, showcasing its significant antiviral potential (Jamison et al., 2025).
  • Selective amidation: HOPO is essential in synthesizing nirmatrelvir. As a coupling agent with EDCI in the amidation step, it enhances efficiency and minimizes racemization, thereby improving yield and purity. This effective application of HOPO supports scalable and sustainable manufacturing of critical pharmacological compounds. (Russell F et. al., 2023).
  • Optical property: HOPO is used in the synthesis of magnesium(II) complexes with heterocyclic ligands, enhancing their photonic properties. It facilitates the amidation reaction that links key fragments, improving synthesis efficiency and contributing to the structural rigidity necessary for effective luminescence in OLED’s. (Shri Bhagwanet. al., 2022).
Integrating 2-Hydroxypyridine N-oxide into your synthetic approaches boosts efficiency, reliability, and innovation, delivering substantial support for research applications and facilitating seamless scalability in manufacturing.

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany


Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class

11 - Combustible Solids

Flash Point (°F)

212.0 °F - closed cup

Flash Point (°C)

100 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library