Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41105500
NACRES:
NA.51
Quality Segment
usage
1000 purifications
sustainability
Sustainability features
concentration
45-55 mg/mL (Magnetic particle)
test parameters
total test time: 5 min
suitability
suitable for nucleic acid purification
storage temp.
2-30°C
General description
Click here to view its DfS scorecard.
The Ultra pure MagPrep® cfDNA Binding Buffer is intended for use within the Ultra pure MagPrep® nucleic acid purification system for manual or high throughput viral nucleic acid isolation from most biological fluids. The Ultra pure MagPrep® nucleic acid purification system is optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. Buffer is quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. Nucleic acid purified using the Ultra pure MagPrep® system is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
Features and Benefits
- Nucleic Acid Isolation
- Manual or high-throughput automated isolations
- RT-qPCR
- Digital PCR
- Biological samples can be used
- Room temperature stable reagents
- DNA free
- DNase free
- RNase free
- NICKase free
- simple and fast protocols
Legal Information
MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Only
Product No.
Description
- UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles
- UPMPCFDBB-50ML-KC Ultra pure MagPrep cfDNA Binding Buffer
- UPMPWB1-100ML-KC Ultra pure MagPrep Wash Buffer 1
- UPMPWB2-100ML-KC Ultra pure MagPrep Wash Buffer 2
- UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 2 Oral
Target Organs
Thyroid
Storage Class
3 - Flammable liquids
Flash Point (°F)
-10.3 °F
Flash Point (°C)
-23.5 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


