Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.71
UNSPSC Code:
12352200
form
liquid
shelf life
1 yr
manufacturer/tradename
Cell Applications, Inc
packaging
pkg of
storage condition
protect from light
shipped in
wet ice
storage temp.
2-8°C
General description
Basal medium (contains no growth supplement) for keratinocytes and epithelial cells. Add GS before use.
Preparation Note
contained in protocol
Analysis Note
Each lot was tested for the ability to support plating, spreading and proliferation
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.