Skip to Content
MilliporeSigma

939072

2-Bromo-9,9-di(methyl-D3)-9H-Fluorene

≥98 atom % (D)

Synonym(s):

2-Bromo-9,9-di(methyl-D3)-9H-Fluorene, 2-Bromo-9,9-dimethylfluorene, 9,9-Dimethyl-2-bromofluorene, 9H-Fluorene, 2-bromo-9,9-dimethyl

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.11
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


isotopic purity

≥98 atom % (D)

Quality Segment

assay

≥98% (CP)

form

powder (or crystal)

storage temp.

room temp

SMILES string

CC1(C2=CC=CC=C2C3=C1C=C(C=C3)Br)C

InChI

InChI=1S/C15H13Br/c1-15(2)13-6-4-3-5-11(13)12-8-7-10(16)9-14(12)15/h3-9H,1-2H3

InChI key

MBHPOBSZPYEADG-UHFFFAOYSA-N

General description

2-Bromo-9,9-dimethyl-d6 fluorene ≥98 atom % (D) is a high-purity isotope product with a unique chemical profile that belongs to the class of isotopes. Produced by Sigma-Aldrich, we are committed to providing quality isotope products and a reliable supply chain. Available in industrial and pre-pack quantities.

Application

- Analytical method development
- reaction mechanism investigations
- Mass spectrometry calibration
- Pharmaceutical formulation development

Features and Benefits

Features
- Excellent solvent characteristics for NMR applications
- High deuterium content for effective labeling
- Improves the reliability of spectroscopic data
- Non-toxic and safe for laboratory environments

Benefits
- Accelerates drug formulation processes and testing
- Improves the quality of NMR results and analyses
- Increases efficiency in research studies
- Supports the development of new therapeutic strategies


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library