Skip to Content
MilliporeSigma

A9603

Anti-Aph-1aL antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 30 kDa

species reactivity

human

technique(s)

western blot: 2-4 μg/mL using whole extract of human kidney 293 cells transfected with human Aph-1aL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... APH1A(51107)

General description

Aph-1aL is a γ secretase subunit that facilitates the cleavage of several integral membrane proteins such as β-APP and Notch. Aph-1aL abnormalities have been associated with the risk of Alzheimer′s disease.
Rabbit Anti-Aph-1aL antibody recognizes Aph-1aL (30 kDa). Additional bands at approx. 46kDa and 20kDa may be observed in some cell extracts, which may represent Aph-1aL aggregation and degradation products, respectively.
This sequence is identical in mouse Aph-1aL and not found in Aph-1aS and Aph-1b.

Immunogen

synthetic peptide corresponding to amino acids 246-265 located at the C-terminus of human Aph-1aL.

Application

Rabbit Anti-Aph-1aL antibody has been used for western blot applications.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library