Skip to Content
MilliporeSigma

HPA004174

Anti-RARB antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

enhanced validation

Enhanced Validation

Synonym(s):

Anti-HBV-activated protein antibody produced in rabbit, Anti-RAR-β antibody produced in rabbit, Anti-RAR-ε antibody produced in rabbit, Anti-Retinoic acid receptor β antibody produced in rabbit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Human Protein Atlas Number:
UNSPSC Code:
12352203
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IF, IHC
Citations:
-
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:500-1:1000

immunogen sequence

VLSVSPGQILDFYTASPSSCMLQEKALKACFSGLTQTEWQHRHTAQSIETQSTSSEELVPSPPSPLPPPRVYKPCFVCQDKSSGYHYGV

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RARB(5915)

General description

RARB exist with three major isotypes, termed α, β and γ. It is involved in the hormone-independent transcriptional activation. It is expressed in the cytoplasm and sub-nuclear space that resemble nuclear bodies.
The retinoic acid receptor-β(RARB) gene is mapped to human chromosome 3p24.

Immunogen

Retinoic acid receptor β recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-RARB antibody produced in rabbit has been used in immunohistochemical staining.

Biochem/physiol Actions

RARB is a tumor suppressor gene. It interacts weakly with silencing mediator of retinoic acid and thyroid hormone receptor (SMRT) corepressor protein. It plays a pivotal role in mediating the retinoid signaling during morphogenesis, development, and cell differentiation. RARB plays an important role in retinoic acid pathway during eye development and organogenesis. Any recessive and dominant mutation in RARB causes anophthalmia and/or microphthalmia and diaphragmatic hernia. The suppression of RARB by DNA methylation and EZH2 leads to the gene silencing during prostate tumorigenesis.
The retinoic acid receptor-β(RARB) may act as an important target in cancer research. It is involved in the development of cancer and is linked with various human cancer types.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST84809

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library