Skip to Content
MilliporeSigma

HPA022479

Anti-SPAG5 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

enhanced validation

Enhanced Validation

Synonym(s):

Anti-Astrin, Anti-Deepest, Anti-MAP126, Anti-Mitotic spindle-associated protein p126, Anti-Sperm-associated antigen 5

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunohistochemistry: 1:50-1:200

immunogen sequence

KSGELISLREEVTHLTRSLRRAETETKVLQEALAGQLDSNCQPMATNWIQEKVWLSQEVDKLRVMFLEMKNEKEKLMIKFQSHRNILEENLRRSDKELEKLDDIVQHIYKTLLSIPEVVRGCKELQGLLEF

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... SPAG5(10615)

General description

SPAG5 (Sperm associated antigen 5) is a mitotic microtubule-associated protein highly expressed in testis of pachytene spermatocytes.

Immunogen

Sperm-associated antigen 5 recombinant protein epitope signature tag (PrEST)

Application

Anti-SPAG5 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

SPAG5 (Sperm associated antigen 5) plays a vital role in the mitosis and cell cycle regulation by controlling the formation and organization of microtubule, checkpoint regulation during cell division. It acts as a regulator and binds to the spindle during separation of sister chromatids. It has also been reported that similar to other cell cycle-related proteins, it also perform a role in the DNA damage repair. It has been suggested that overexpression of SPAG5 may indicate a pathogenic role in testicular development of patients with Down′s syndrome and cryptorchidism.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Other Notes

Corresponding Antigen APREST75532.

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

— or —

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library