Skip to Content

N8283

Monoclonal Anti-Nicotinic Acetylcholine Receptor (β1 Subunit) antibody produced in rat

~5 mg/mL, clone mAb 111, purified immunoglobulin, buffered aqueous solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352203
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rat

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

mAb 111, monoclonal

form

buffered aqueous solution

species reactivity

eel, rat, human, mouse

concentration

~5 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable, immunoprecipitation (IP): suitable, western blot: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... CHRNB1(1140)
mouse ... Acp5(11433)
rat ... Chrnb1(24261)

General description

Cholinergic receptor nicotinic β 1 subunit (CHRNB1) is predominantly expressed in muscles. It is also reported to be expressed in the brain, lungs and adrenal glands. The gene encoding this protein is localized on human chromosome 17p13.1.

Immunogen

denatured nicotinic acetylcholine receptors from the electric eel Torpedo.

Application

Monoclonal Anti-Nicotinic Acetylcholine Receptor (β1 subunit) antibody produced in rat has been used in immunohistochemistry.

Biochem/physiol Actions

Lack of expression of cholinergic receptor nicotinic β 1 subunit (CHRNB1) has been linked to weak channel activity post agonist binding. Mutation in the gene encoding the protein has been associated with congenital myasthenic syndrome.
Recognizes an epitope within amino acids 360-410 of the muscle nicotinic acetylcholine receptor, β1 subunit. Binds native and denatured β1.

Physical form

Solution in 10 mM phosphate buffered saline, containing 10 mM sodium azide.

Legal Information

Sold with the permission of the Salk Institute

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library