Skip to Content
MilliporeSigma

ABK0019

p53Bridge ELISA

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
41116158
Species reactivity:
human, mouse
Storage temp.:
2-8°C
Shipped in:
wet ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


antibody form

purified antibody

Quality Segment

species reactivity

human, mouse

technique(s)

ELISA: suitable

shipped in

wet ice

storage temp.

2-8°C

General description

A Protein Conformational Array (PCA) in a sandwich ELISA format where the plate is coated with a panel of 17 antibodies raised against peptides derived from the full length protein sequence of p53. Taken individually, each of these antibodies is strongly antigenic to the peptide sequence that was used in its production. However, when these peptides are incorporated into a full length correctly folded protein, the antigenicity of some of them is masked by the three dimensional structure of the protein and only a certain number of the antibodies respond. The result is a histogram which can be likened to a ‘fingerprint′ for correctly folded p53. For a testing sample from cancer tissue or cell line, if the protein is not mutated or the mutations do not impact the p53 HOS, the ‘fingerprint′ will match that of wild type p53. If the p53 mutations resulted in the alteration of the protein HOS, changes in the ‘fingerprint′ generated by the ELISA will point out differences between the mutant and wild type p53, this information may be valuable in the characterization of the p53 from cancer patient samples, cancer cell lines or other biological samples.

Application

For research use only. Not for use in diagnostic procedures.

Other Notes

ELISA kit contains three 96-well microplates and all the components necessary to complete an ELISA test.


Pictograms

Corrosion

Signal Word

Warning

Hazard Codes

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library