Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12352202
eCl@ss:
32039090
recombinant
expressed in E. coli
assay
>80% (SDS-PAGE)
form
buffered aqueous solution
mol wt
predicted mol wt 32 kDa
purified by
immobilized metal affinity chromatography (IMAC)
concentration
≥0.5 mg/mL
immunogen sequence
PVSGDLESNPDYWSGVMGGSSGASRGPSAMEYNKEDRESFRHSQPRSKSEMLSRKNFATGVPAVSMDELAAFADSYGQRPRRADGNSHEARGGSRFERSESRAHSGFYQDDSLEEYYGQRSRSREPLTDADR
Ensembl | human accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... ILDR2(387597)
General description
Recombinant protein fragment of Human ILDR2 with N-terminal His6ABP (ABP = Albumin Binding Protein derived from Streptococcal Protein G) fusion tag.
Application
Suitable as a blocking agent using corresponding antibodies.
Physical form
Solution in 1 M urea-PBS, pH 7.4
Preparation Note
The protein solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user.
Other Notes
Corresponding Antibody HPA012545.
Legal Information
Prestige Antigens is a trademark of Sigma-Aldrich Co. LLC
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class
10 - Combustible liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.