Skip to Content
MilliporeSigma

AXYLT20RL

Corning® Axygen® LTS Compatible Pipet Tip

maximum volume 20 μL, Reload Stack

Synonym(s):

Pipette tips, sustainable pipet tip, sustainable pipette tip

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
Sterility:
non-sterile
Feature:
DNA free (Human), PCR clean, RNase and DNase free, autoclavable, graduations, pyrogen free
Packaging:
pkg of 4 packs of 960
Manufacturer/tradename:
Corning LT-20-RL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

non-sterile

feature

DNA free (Human), PCR clean, RNase and DNase free, autoclavable, graduations, pyrogen free

manufacturer/tradename

Corning LT-20-RL

packaging

pkg of 4 packs of 960

maximum volume

20 μL

General description

The Axygen LTS compatible tips are ergonomically designed to reduce discomfort and optimized to work with Rainin LiteTouch Pipettors, all with the quality you have come to expect from Corning. The tips offer comfort you can feel with a reduction of force required to seal and eject, enabling a streamlined experience from aspiration to dispensing. Avoid slamming with excessive force on the pipettor for a proper seal, eliminate the struggle to eject tips, and minimize sore hands and wrists after extensive pipetting. Tips are offered in a variety of packaging formats; racked, racked/filtered, tip station, reload stacks, and our more sustainable solution: Axygen HybridRack™ Pipet Tips. Axygen LTS compatible tips are manufactured in the USA and meet ISO standards. All tips are certified RNase-/DNase-free, Human gDNA-, PCR-inhibition-free, and nonpyrogenic.

Application

Pipet tips are used across a wide variety of applications including molecular biology, microbiology, cell culture, clinical diagnostics, chemical analysis, and protein biochemistry. Specific workflows include: PCR (polymerase chain reaction), DNA sequencing, DNA purification, RNA isolation, cloning, gene expression analysis, inoculating cultures, performing microbial assays, preparing bacterial fungal cultures, seeding cells, transferring media and reagents, performing cell based assays, handling cell cultures in plates or flasks, sample preparation, liquid transfers in diagnostic assays, various medical testing procedures that require precise volumes, dispensing reagents, preparing solutions, and protein purification/quantification.

Features and Benefits

  • Flexible sealing area optimized to work with LTS pipettors enables less force to be used during pipetting
  • Graduation lines for quick accuracy checks
  • Fine point design for crisp sample dispensing
  • Ultra Clear USP Class VI resin
  • Non-sterile product is autoclavable at 122°C for 15 minutes

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library