Skip to Content
MilliporeSigma

AXYMRF10XTRS

Corning® Axygen® MultiRack Extended Length Filtered Tip

10uL, 4800 tips

Synonym(s):

pipet tip, pipette tip, pipettor tips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121600
Volume:
10 μL
Sterility:
sterile
Feature:
autoclavable: no
Packaging:
pack of 5 packs of 960 tips
Manufacturer/tradename:
Corning MRF-10XT-R-S
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

sterile

feature

autoclavable: no

manufacturer/tradename

Corning MRF-10XT-R-S

packaging

pack of 5 packs of 960 tips

volume

10 μL

General description

Axygen® universal pipet tips are designed to fit a wide variety ofsingle- and multi-channel pipettors. Flexible walls and a series ofinternal sealing rings ensure a secure fit with less force requiredto load and eject.The Axygen product offering now also includes the AxygenMultiRack System, which is our updated reload and rackingsystem. In addition to the universal fit tips packaged in theMultiRack System, we now also offer LTS Compatible tips in theMultiRack System, and if you are worried about your carbonfootprint, the Axygen HybridRack™ is a perfect option for you.

Application

liquid handling

Features and Benefits

  • Automated assembly – Ensures product is free from contamination.
  • ISO 13485 and ISO 14001 – Certified quality management systems supply stringent quality processes to ensure the highest quality finished good.
  • In-house testing – In-house QA lab allows product to be validated for fit, form, and function prior to release.
  • Brand you can trust – A long track record of innovation and value as one of the Corning trusted brands.
  • Broad product offering – Corning offers many different styles of pipet tips, including racked, filtered, reloads, stack racks, low retention, wide bore, and more. With so many options, you are sure to find a tip that works for you.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
HybridRack is a trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library