Skip to Content
MilliporeSigma

AXYVT255R

Corning® Axygen® 96-well tips

250µL, non-sterile, 4800 tips

Synonym(s):

pipet tip, pipette tip, pipettor tips

Select a Size

Change View

About This Item

UNSPSC Code:
41121600
eCl@ss:
32040201
Volume:
250 μL
Sterility:
non-sterile
Feature:
autoclavable: no
Packaging:
pack of 4800 tips
Manufacturer/tradename:
Corning VT-255-R
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

non-sterile

feature

autoclavable: no

manufacturer/tradename

Corning VT-255-R

packaging

pack of 4800 tips

volume

250 μL

General description

Axygen® universal pipet tips are designed to fit a wide variety ofsingle- and multi-channel pipettors. Flexible walls and a series ofinternal sealing rings ensure a secure fit with less force requiredto load and eject.The Axygen product offering now also includes the AxygenMultiRack System, which is our updated reload and rackingsystem. In addition to the universal fit tips packaged in theMultiRack System, we now also offer LTS Compatible tips in theMultiRack System, and if you are worried about your carbonfootprint, the Axygen HybridRack™ is a perfect option for you.

Application

liquid handling

Features and Benefits

  • Automated assembly – Ensures product is free from contamination.
  • ISO 13485 and ISO 14001 – Certified quality management systems supply stringent quality processes to ensure the highest quality finished good.
  • In-house testing – In-house QA lab allows product to be validated for fit, form, and function prior to release.
  • Brand you can trust – A long track record of innovation and value as one of the Corning trusted brands.
  • Broad product offering – Corning offers many different styles of pipet tips, including racked, filtered, reloads, stack racks, low retention, wide bore, and more. With so many options, you are sure to find a tip that works for you.

Legal Information

Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
HybridRack is a trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Axygen® 96-well tips




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library