Skip to Content
MilliporeSigma

BR26547

BRAND®, accu-jet® S snap tape

polyolefin, pkg of 3 ea

Synonym(s):

Accu-Jet® S accessories, Pipette controller snap tape

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.04
UNSPSC Code:
41121514
eCl@ss:
32070201
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyolefin

packaging

pkg of 3 ea

manufacturer/tradename

BrandTech 26539

Application

Accessories for BRAND®, accu-jet® S pipette controller

Legal Information

BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library