Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
SB.56
UNSPSC Code:
41121514
material
polypropylene
packaging
pkg of 1 ea
manufacturer/tradename
BRAND 26658
color
white
General description
Accessories for BRAND®, accu-jet® S pipette controller
Application
holding silicone adapter of accu-jet® S
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
accu-jet is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.