Skip to Content
MilliporeSigma

CAS9CBEPRC

PURedit® Cas9 Precision Cytosine Base Editor

Engineered, Recombinant, expressed in E. coli

Synonym(s):

CBE, Cas9 Cytosine Base Editor

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116134
NACRES:
NE.03
Concentration:
8 mg/mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in E. coli

Quality Segment

form

solution (50% glycerol)

shelf life

2 yr

mol wt

214 kDa

concentration

8 mg/mL

application(s)

CRISPR

shipped in

dry ice

storage temp.

-65 to -96°C

General description

PURedit® Cas9 Precision Cytosine Base Editor has been engineered to achieve defined nucleotide changes of cytosine to thymine using RNP complex delivery. The PURedit® Cas9 Precision Cytosine Base Editor will edit cytosine residues from the third position of the protospacer to the ninth position. The base editor protein is recombinantly expressed in E. coli with 2X NLS. Uracil glycosylase inhibitor (UGI) protein, also recombinantly expressed in E. coli with 2X NLS, is included as a recommended optional component to enhance the rate of C-to-T editing and reduce the rate of other editing outcomes, including insertions and deletions (indels). UGI may be omitted to enable lower-frequency C-to-R (R = A or G) editing..
The kit contains:
  • Cytosine Base Editor protein (CAS9CBEPRC) in 50% glycerol at 8 mg/mL
  • Uracil glycosylase inhibitor (UGIPRT) in 50% glycerol at 15 mg/mL
  • Transfection buffer (PEXBUF)
  • Dilution buffer (DBUFFER)

For best performance, gRNA sequence should be designed using our free online DNA base editor tool. Link here: SigmaAldrich.com/baseeditordesign

We also recommend our gRNA products for use with the PURedit® Cas9 Cytosine Base Editor kits. Order your custom gRNA here: SigmaAldrich.com/customgrna

Application

  • Genome editing
  • Functional genomics
  • CRISPR screening
  • Cell culture
  • Embryos

Features and Benefits

Best in class protein offering of cytosine base editors. Safer gene editing without double stranded breaks allows for editing in sensitive cell lines and eliminates the risk of nonspecific donor DNA integration.

Precision Editing: high rates of C-to-T substitution at only the target site; reduced risks of off-target edits relative to plasmid or viral delivery.

Resource savings: screen fewer clones due to more predictable editing.

Other Notes

Please see the following reference for more understanding of the development and characterization process: https://link.springer.com/article/10.1186/s12896-025-01020-1

Legal Information

PURedit is a registered trademark of Merck KGaA, Darmstadt, Germany


Storage Class

10 - Combustible liquids

WGK

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library