Skip to Content
MilliporeSigma

CLS357474

Corning® Silicone pipette holder

case of 1

Synonym(s):

pipet accessory, pipette accessory

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121517
eCl@ss:
32160703
Material:
silicone
Compatibility:
Falcon® pipet controller
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

silicone

feature

autoclavable

packaging

case of 1

manufacturer/tradename

Corning 357474

compatibility

Falcon® pipet controller

General description

Replacement silicone holder for Falcon® pipet controller.

Application

Cell culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Falcon is a registered trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library