Skip to Content
MilliporeSigma

CLS3763

Corning® Microplate

384 Well, Polystyrene, White/Clear Flat Bottom, Square Wells, Not Treated, no Lid, Nonsterile, Bulk

Synonym(s):

multiwell plate, MTP plate, assay plate, microtiter plate, microwell plate, well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41113036
Material:
polystyrene
Sterility:
non-sterile
Binding type:
non-treated surface
Feature:
lid: no, wells 384
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

non-sterile

feature

lid: no, wells 384

packaging

case of 100 (4 packs of 25)

manufacturer/tradename

Corning 3763

plate wells

112 μL

plate working volume

20 μL (80 μL)

binding type

non-treated surface

General description

  • 25 per Bag, without Lid, Nonsterile
  • Ideally suited for luminescent assays using either top or bottom detection plate readers
  • Recommended working volume of up to 20 to 80 µL
  • Conforms to standard microplate footprint and dimensions
  • Not treated (or medium binding) polystyrene surface is hydrophobic in nature and binds biomolecules through passive interactions
  • Suitable primarily for the immobilization of large molecules, such as antibodies, that have large hydrophobic regions that can interact with the surface
  • Untreated microplates have a binding capacity of approximately 100 to 200ng IgG/cm²

Application

assay plate

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Microplate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library