Skip to Content
MilliporeSigma

CLS3767

Corning® Microplate

384 Well, Polystyrene, White/Clear Flat Bottom, Square Wells, Non Binding Surface, no Lid, Nonsterile, Bulk

Synonym(s):

MTP plate, assay plate, microtiter plate, microwell plate, multiwell plate, well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41113036
Material:
polystyrene
Sterility:
non-sterile
Binding type:
NBS™ (non-binding surface)
Feature:
lid: no, wells 384
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

non-sterile

feature

lid: no, wells 384

packaging

case of 100 (4 packs of 25)

manufacturer/tradename

Corning 3767

plate wells

112 μL

plate working volume

20 μL (80 μL)

binding type

NBS™ (non-binding surface)

General description

  • 25 per Bag, without lid, nonsterile
  • White walled plates enhance luminescent signals and have low background luminescence and fluorescence
  • Nonbinding (NBS) surface is a Corning proprietary treatment technology used on polystyrene microplates to create a nonionic hydrophilic surface (polyethylene oxide-like) that minimizes molecular interactions Ideal for reducing protein and nucleic acid binding at low concentrations, and increasing assay signal to noise Ideally suited for luminescent assays using either top or bottom detection plate readers
  • Recommended working volume of up to 20 to 80 µL
  • Conforms to standard microplate footprint and dimensions

Application

assay plate

Legal Information

Corning is a registered trademark of Corning, Inc.
NBS is a trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Microplate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library