Skip to Content
MilliporeSigma

CLS400640

Corning® Reusable Spinner Direct Top Drive Motor

AC/DC input 120 V AC, 60 Hz

Synonym(s):

equipment accesory, repair part

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41103025
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


AC/DC input

120 V AC

material

aluminum

sterility

non-sterile

feature

reusable

packaging

case of 1

manufacturer/tradename

Corning 400640

application(s)

cell culture

General description

High torque, low rpm stirrer motor specifically designed to maintain constant low speed for use on top of Corning® Reusable Spinner Flasks. The gearhead stirrer motor delivers 14.5 in-lbs of torque and has a maximum speed of 350 rpm. The weight of the motor is 9 lbs (4.1kg) and operates on 120VAC 60Hz.

Application

Cell culture

Legal Information

Corning is a registered trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library