Skip to Content
MilliporeSigma

CLS4445

Corning® Elplasia Plate

24 well, Black/Clear, Square Bottom, Plasma-treated Microcavity Plate with Lid, Sterile, Individually Packaged

Synonym(s):

24 well plate, cell culture plate, microplate, plate, spheroid plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122102
NACRES:
NB.02
Material:
black polystyrene , square wells
Sterility:
sterile
Binding type:
(plasma treated)
Feature:
lid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

black polystyrene , square wells

sterility

sterile

feature

lid

packaging

case of 5

manufacturer/tradename

Corning 4445

plate wells

3.47 mL

plate working volume

2 mL

binding type

(plasma treated)

General description

The only plates with a simple “plug and play” protocol for scaffold-free, self-assembly spheroid formation at large volumes. Corning Elplasia square bottom plates feature a surface with optical qualities suited for image analysis, making them an ideal solution for clonal selection and high magnification imaging of very small clusters. The Elplasia square plate is well suited for screening programs as it generates a high volume of spheroids per well, allowing for higher signals per well. Square bottom plates are plasma-treated for self-coating and come in 6-, 24-, 96- and 384-well formats.

Application

Spheroid plate

Features and Benefits

  • Spheroids may be formed and cultured for 21 or more days
  • Can produce between 79 – 15,000+ spheroids per well, depending on plate format, under one culture condition
  • The high volume of spheroids generated in each well increase signals per well without increasing spheroid size
  • Black opaque sidewalls to reduce well-to-well “cross-talk”
  • Well suited for luorescent/luminescent assays
  • Available in multiple formats, two well geometries and surface coating options.

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Elplasia Plate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library