Skip to Content
MilliporeSigma

CLS4588

Corning® Microplate

384 Well, Polystyrene, Black Clear Bottom, Flat Bottom, ULA coated,w/lid, Sterile, Bulk 20/100

Synonym(s):

MTP plate, assay plate, microtiter plate, microwell plate, multiwell plate, well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41113036
Material:
polystyrene
Sterility:
sterile
Binding type:
low binding surface
Feature:
lid, wells 384
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

sterile

feature

lid, wells 384

packaging

case of 100 (5 packs of 20)

manufacturer/tradename

Corning 4588

plate wells

112 μL

plate working volume

20 μL (80 μL)

binding type

low binding surface

General description

  • 20 per Bag, with Lid, Sterile
  • Ultra-Low Attachment microplates feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation and cellular activation
  • Surface is noncytotoxic, biologically inert and nondegradable by gamma irradiation and nonpyrogenic
  • Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Can be used for both top and bottom reading instruments
  • Bulk packed 20 per bag

Application

assay plate

Legal Information

Corning is a registered trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Microplate




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library