Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122101
NACRES:
NB.02
Material:
flat wells, polystyrene
Sterility:
sterile
Binding type:
(ultra-low attachment)
Feature:
lid
material
flat wells, polystyrene
sterility
sterile
feature
lid
packaging
case of 40, pack of 5
manufacturer/tradename
Corning 4615
plate wells
16.5 mL
plate working volume
11 mL (16.5 mL)
color
clear
binding type
(ultra-low attachment)
General description
Ultra-Low Attachment dishes feature a covalently bound hydrogel layer that effectively inhibits cellular attachment
Application
cell culture
Features and Benefits
- Approximate growth surface area is 55 cm²
- Surface minimizes protein absorption, enzyme activation, and cellular activation
- Surface is non-cytotoxic, biologically inert, and nondegradable
- Sterilized by gamma radiation and nonpyrogenic
- Actual inside growth surface diameter is 83.8mm
- Height is 20mm
- Manufactured from optically-clear virgin polystyrene
- Have stacking beads to aid in handling
- Supplied with vents to provide consistent gas exchange
Legal Information
Corning is a registered trademark of Corning, Inc.
Still not finding the right product?
Explore all of our products under Corning® Ultra-low attachment culture dishes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.