Skip to Content
MilliporeSigma

CLS72592

Corning® Costar® Stripwell™ Microplate

96 Well Stripwell™, 8 Well Strips, Polystyrene, Clear, Flat Bottom, High Binding, no Lid, Nonsterile, Bulk, Special Package

Synonym(s):

MTP plate, assay plate, microtiter plate, microwell plate, multiwell plate, well plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41113036
Material:
flat bottom clear polystyrene
Sterility:
non-sterile
Binding type:
high binding surface
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

flat bottom clear polystyrene

sterility

non-sterile

feature

lid: no

packaging

case of 100 (4 packs of 25)

manufacturer/tradename

Corning 72592

plate wells

360 μL

plate working volume

75 μL (200 μL)

binding type

high binding surface

Application

assay plate

Legal Information

Corning is a registered trademark of Corning, Inc.
Costar is a registered trademark of Corning, Inc.
Stripwell is a trademark of Corning, Inc.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library