Skip to Content
MilliporeSigma

CLSSA0910

Corning® Gosselin™ Autoclavable Bag

capacity 9 L, Black Biohazard Logo

Synonym(s):

Autoclave Bag, Heat-Resistant Bag, Sterilization Bag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
24111500
Material:
polypropylene
Sterility:
non-sterile
Capacity:
9 L
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene

sterility

non-sterile

feature

Black Biohazard Logo

packaging

case of 500

manufacturer/tradename

Corning SA09-10

capacity

9 L

General description

Autoclavable bags designed for disposal of laboratory waste, offering high-temperature resistance and reliable containment.

Application

Environmental and Water Testing, Food and Beverage Testing, Life Science

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library