Skip to Content
MilliporeSigma

CLSSB93101

Corning® Gosselin™ Petri Dish

capacity 100 mm × 15 mm, 3 Vents

Synonym(s):

Agar Plate, Culture Plate, Petri Plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122101
eCl@ss:
32040503
Material:
polystyrene
Sterility:
non-sterile
Manufacturer/tradename:
Corning SB93-101
Packaging:
case of 825
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polystyrene

sterility

non-sterile

feature

3 Vents

packaging

case of 825

manufacturer/tradename

Corning SB93-101

capacity

100 mm × 15 mm

General description

High-quality polystyrene petri dishes engineered for microbiology applications, offering excellent clarity, controlled venting, and reliable culture growth conditions.

Application

Microbiology / Culture

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Gosselin Petri Dish




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library