Skip to Content
MilliporeSigma

CLSSM101

Corning® Gosselin™ Blender Bag

capacity 80 mL, No Filter, sterile

Synonym(s):

Homogenizer Bag, Sample Mixing Bag, Stomacher Bag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41101518
Material:
polyethylene
Sterility:
sterile
Capacity:
80 mL
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polyethylene

sterility

sterile

feature

No Filter

packaging

case of 4500

manufacturer/tradename

Corning SM1-01

capacity

80 mL

General description

Sterile polyethylene blender bags for sample homogenization and preparation, available with or without filters to support microbiological testing workflows.

Application

Sample Preparation / Homogenization

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.


Still not finding the right product?

Explore all of our products under Corning® Gosselin Blender Bag




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library