Skip to Content
MilliporeSigma

CLSTP53C002

Corning® Gosselin™ Straight Container

capacity 180 mL, red screw cap

Synonym(s):

Rigid Container, Sample Container, Straight-Sided Container

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41123300
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

polypropylene , red screw cap

sterility

non-sterile

feature

cap

packaging

case of 264

manufacturer/tradename

Corning TP53C-002

capacity

180 mL

General description

Polypropylene straight containers designed for secure sample collection and storage, featuring durable construction and various cap options for easy handling.

Application

Sample Collection & Storage

Legal Information

Corning is a registered trademark of Corning, Inc.
Gosselin is a trademark of Corning, Inc.


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library