Skip to Content
MilliporeSigma

DWK953901-2002

KIMBLE® ULTRA-WARE HPLC Reservoir

capacity 2 L, conical bottom plastic-coated glass, pack of 1 ea

Synonym(s):

953901-2002, HPLC reservoir, KIMBLE ULTRA-WARE reservoir, ungraduated reservoir

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
24111800
NACRES:
NB.43
Manufacturer/tradename:
DWK Life Sciences Kimble® 953901-2002
Capacity:
2 L
Material:
conical bottom plastic-coated glass
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

conical bottom plastic-coated glass

feature

graduation: no

packaging

pack of 1 ea

manufacturer/tradename

DWK Life Sciences Kimble® 953901-2002

parameter

-1—0.4 bar pressure

H

319 mm

capacity

2 L

compatibility

made to hold HPLC reagents

General description

Designed for preparation, storage and delivery of all liquid chromatography mobile phases. As part of our ULTRA-WARE series, these reservoirs are recommended for use under vacuum for filtration and vacuum degassing of mobile phase solvents. Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.

Application

storage, reagent prep

Features and Benefits

  • Conical bottom delivers all of the mobile phase without reservoir tilting
  • Side neck style allows addition of filtered solvent or pH meter without removing delivery cap
  • Plastic (PVC) safety-coating blocks UV light up to 385 nm, preventing photo-degradation of light-sensitive reagents
  • Safety coating helps to retain glass fragments and allows a reasonable amount of time for the safe disposal of liquid contents
  • Used with internal pressures up to 0.4 bar (6 psig) for helium sparging and blanketing of the mobile phase
  • Operating Pressure: -1.0 to 0.4 bar (-14.5 psig to 6 psig)Chemically inert glass prevents leaching of any extractables into the mobile phase solvents
  • Supplied with GL 45 screw thread

Legal Information

KIMBLE is a registered trademark of DWK Life Sciences


Still not finding the right product?

Explore all of our products under KIMBLE® ULTRA-WARE HPLC Reservoir




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library