Skip to Content
EP0030113624
Eppendorf® Twin tec PCR plates, 96 wells
skirted 2D SafeCode, colorless wells, blue Plate, pack of 25 plates (5 bags × 5 plates)
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41106300
Sterility:
non-sterile
Feature:
PCR clean, skirt
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
sterility
non-sterile
feature
PCR clean, skirt
packaging
pack of 25 plates (5 bags × 5 plates)
manufacturer/tradename
Eppendorf® 0030113624
well maximum volume
150 μm
well size
96 wells
color
blue Plate, colorless wells
General description
Choose Eppendorf twin.tec PCR plates for confidence in every PCR assay. twin.tec plates offer purity, rigidity, color, and other simple, smart features can make your daily life a lot easier. The rigid polycarbonate frame works well in automation and high-throughput processes, and the different colors mean you can quickly organize the different steps of your workflow.
Legal Information
Eppendorf is a registered trademark of Eppendorf SE
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)