Skip to Content
MilliporeSigma

LGC600524

LGC E.Cloni 10G Elite Electrocompetent Cells

pkg of 48 reactions (8 x 150 µL), Cfu/μg >2 x 1010

Synonym(s):

High Efficiency competent cells, DNA transformation, Electrocompetent E.Coli, Electroporation, competent cells

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106202
NACRES:
NB.43
Biological source:
Escherichia coli
Growth mode:
growth or suspension
Manufacturer/tradename:
Lucigen 60052-4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Escherichia coli

manufacturer/tradename

Lucigen 60052-4

packaging

pkg of 48 reactions (8 x 150 µL)

growth mode

growth or suspension

technique(s)

microbiological culture: suitable

cell transformation

competent cell type: electrocompetent
transformation efficiency: 2 x 1010 cfu/μg

shipped in

dry ice

storage temp.

-65 to -96°C

General description

E. cloni electrocompetent cells incorporate a unique manufacturing technology that increases transformation efficiency, recombinant yields and reliability. These cells provide solutions for a wide range of applications at economical prices.

Application

E. cloni 10G Competent Cells. Library construction, cloning, subcloning, and plasmid isolation with or without blue/white screening.

Features and Benefits

  • Optimized genetics for high yields: phage T1 resistant, endonuclease and recombination minus, blue/white screening-capable (without IPTG induction)
  • Supports cloning methylated DNA
  • Replace standard cloning strains (e.g., OP10, DH5α, DH10B, XL1-Blue, etc.)

Transformation efficiency: ≥ 2 x 1010 (cfu/µg pUC DNA)

Genotype: E. cloni 10G: F¯ mcrA Δ(mrr-hsdRMS-mcrBC) endA1 recA1 ɸ80dlacZΔM15 ΔlacX74 araD139 Δ(ara,leu)7697 galU galK rpsL (StrR) nupG λ- tonA

Components:


E. cloni 10G Elite Electrocompetent Cells


pUC 19 Transformation Control


Recovery Media


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library