Skip to Content
MilliporeSigma

M2772

7-Methyl-2′-deoxyguanosine 5′-triphosphate sodium salt

~90%

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C11H18N5O13P3
CAS Number:
Molecular Weight:
521.21
UNSPSC Code:
12352204
MDL number:
eCl@ss:
32150313
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

~90%

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].C[n+]1cn([C@H]2C[C@H](O)[C@@H](COP([O-])(=O)OP([O-])(=O)OP([O-])([O-])=O)O2)c3N=C(N)NC(=O)c13



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library