Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Linear Formula:
CH3OC6H4CH2CO2H
CAS Number:
Molecular Weight:
166.17
EC Number:
217-282-8
UNSPSC Code:
12352100
NACRES:
NA.22
Beilstein/REAXYS Number:
2614004
MDL number:
mp
65-69 °C (lit.)
SMILES string
COc1cccc(CC(O)=O)c1
InChI
1S/C9H10O3/c1-12-8-4-2-3-7(5-8)6-9(10)11/h2-5H,6H2,1H3,(H,10,11)
InChI key
LEGPZHPSIPPYIO-UHFFFAOYSA-N
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.