Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41116104
NACRES:
NA.21
eCl@ss:
40050204
usage
sufficient for 100 colorimetric or fluorometric tests
Quality Segment
application(s)
advanced drug delivery
detection method
colorimetric, fluorometric
relevant disease(s)
diabetes; obesity; endocrinological disorders; endocrinological disorders
shipped in
dry ice
storage temp.
−20°C
General description
Glycogen is a branched polysaccharide composed of glucose units, primarily stored in the liver and muscle as a vital short-term energy reserve. Its ability to quickly release glucose to meet sudden energy demands makes it essential for metabolic regulation. Detecting glycogen levels can provide insights into metabolic disorders, such as diabetes and genetic glycogen storage diseases. In diabetes, glycogen metabolism can become disrupted, leading to abnormal accumulation or depletion in the liver due to insulin imbalances. Genetic glycogen storage diseases arise from enzyme deficiencies affecting glycogen synthesis or breakdown.
Application
Suitable for determining glycogen concentration in various tissues, including liver, and in cell culture (both adherent and suspension cells).
The Glycogen Assay Kit has been used to determine glycogen content in liver tissue, as well as in vastus lateralis and muscle homogenates.
Biochem/physiol Actions
Glycogen concentration is determined by a coupled enzyme assay, which produces a colorimetric (570 nm)/ fluorometric (λex = 535/λem = 587 nm) product, proportional to the glycogen present.
Features and Benefits
Compatibility with High-Throughput Systems: Seamlessly integrate our kit into high-throughput handling systems, ensuring smooth operations and accurate results, enhancing efficiency in your laboratory workflow.
Convenient Data Analysis: Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file
Convenient Data Analysis: Supportive calculator will calculate the reagents needed according to your experimental needs, and analyze your results based on your experimental data.
Click here to download a calculator excel file
Still not finding the right product?
Explore all of our products under Glycogen Assay Kit
Signal Word
Danger
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 3 - ED ENV 1
Storage Class
10 - Combustible liquids
WGK
WGK 2
Flash Point (°F)
188.6 °F
Flash Point (°C)
87 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.