Skip to Content
MilliporeSigma

MAK620

Trypsin Inhibitor Assay Kit

sufficient for 100 fluorometric tests

Synonym(s):

Trypsin Inhibitor Screening Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116104
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 fluorometric tests

detection method

fluorometric

storage temp.

−20°C

General description

Trypsin (EC3.4.21.4) is a digestive, serine protease that hydrolyzes dietary proteins in many eukaryotic and prokaryotic organisms. Trypsin predominantly cleaves peptide chains at the carboxyl side of lysine and arginine amino acids, but not before proline.

Application

Suitable for evaluation of drugs and screening of potential inhibitors to trypsin proteases.

Biochem/physiol Actions

The Trypsin Inhibitor Assay Kit uses a fluorescein isothiocyanate (FITC)-labeled synthetic substrate. The fluorescein label is highly quenched. Upon digestion by trypsin present in the sample, the substrate is cleaved into smaller peptides, which abolishes the quenching of the fluorescence label. The fluorescence or fluorescence polarization (FP) of the FITC-labeled fragments is measured at λex/em = 485/530 nm. Inhibition is determined by the decrease in fluorescence.


Storage Class

10 - Combustible liquids

WGK

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library