Skip to Content
MilliporeSigma

MAK628

L-Carnitine Assay Kit

sufficient for 100 colorimetric or fluorometric tests

Synonym(s):

Carnitine Detection Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116104
eCl@ss:
40050204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 colorimetric or fluorometric tests

detection method

colorimetric, fluorometric

storage temp.

−20°C

General description

L-Carnitine is a naturally occurring compound biosynthesized from lysine and methionine that is essential for cellular energy metabolism, with the highest concentrations in skeletal and cardiac muscle. Its primary function is facilitating the transport of long-chain fatty acids into the mitochondrial matrix for β-oxidation via the carnitine shuttle. Beyond transport, L-carnitine serves a critical acyl-group buffering function by reversibly forming acylcarnitines of varying chain lengths, which maintains free CoA availability for essential metabolic reactions, enables export of excess or toxic acyl intermediates, and generates metabolite signatures that reflect the balance between substrate supply, oxidative capacity, and metabolic bottlenecks. Carnitine metabolism is a central hub connecting fatty acid oxidation, mitochondrial function, branched‑chain amino acid catabolism, and cellular energy homeostasis across physiological and disease states.

Application

Suitable for the determination of L-Carnitine in urine, serum, plasma, cell lysates, tissue lysates and food products (e.g. milk).

Biochem/physiol Actions

In this assay, a multistep reaction produces H2O2 which reacts with a specific dye to form a pink colored product. The optical density at 570 nm or fluorescence intensity at λex/em = 530/585 nm is directly proportional to the L‑Carnitine concentration in the sample.

Detection range:

Colorimetric assay 12 ‑ 1000 µM

Fluorometric assay 1 ‑ 100 µM


Storage Class

10 - Combustible liquids

WGK

nwg

Flash Point (°F)

188.6 °F - closed cup

Flash Point (°C)

87 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library