Skip to Content
MilliporeSigma

MTCA8003R

MTC™ Bio BowTie™ Drawstring Biohazard Bags

high-density polyethylene, capacity 33 gal (113 L), W × L 33 in. (84 cm) × 41 in. (104 cm), pack of 100 ea

Synonym(s):

MTC™ Bio BowTie™ Drawstring Biohazard Bags, bio waste bag, waste disposal receptical, MTC Bio A8003R, lab waste bag, waste disposal bag

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
24111500
NACRES:
NB.43
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

high-density polyethylene , red

feature

autoclavable, label with large marking area and sterilization indicator

packaging

pack of 100 ea

manufacturer/tradename

MTC™ Bio A8003R

W × L

33 in. (84 cm) × 41 in. (104 cm)

capacity

33 gal (113 L)

thickness

2.25 mil

General description

The exclusive drawstring loops on BowTie™ Biohazard Bagsprovide instant-cinch closure on your waste materials and a handle to easilyremove bags from their bins. They allow venting during sterilization andprovide an easy, safe way to remove bags from a hot autoclave. Made from 2.25 mil thick HDPE these bags ares suitable for autoclaving in 121°C cycles and are printed with a number of identifyingmarkers such as large biohazard symbols and temperature limit warnings. Theyare also outfitted with a large marking area and a sterilization patch thatchanges colour after autoclaving. BowTie™ Bags fit their respectivebiohazard bins perfectly.

Features and Benefits

  • Cinch closure, no wires needed
  • Colour changing sterilization indicator
  • Biohazardsymbols and temperature limit warnings
  • Allow venting during sterilization
  • Safe& easiy removal from bin or autoclave
  • 2.25mil thick HDPE

Legal Information

BowTie is a trademark of MTC Bio
MTC is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library