Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
Capacity:
5 mL
Material:
clear , conical bottom (self standing), polypropylene , screw cap (separately packed)
Sterility:
sterile
Packaging:
case of 1000 ea (10 bags of 100ea)
Feature:
graduations (molded)
material
clear , conical bottom (self standing), polypropylene , screw cap (separately packed)
description
with marking area
sterility
sterile
feature
graduations (molded)
packaging
case of 1000 ea (10 bags of 100ea)
manufacturer/tradename
MTC™ Bio C1811-SC
capacity
5 mL
diam. × L
16 mm × 80 mm
General description
Used to store, transport or freeze samples, these tubes come in both 5mL (16 x 60mm) and 10mL (16 x 80mm) and come with leak-proof screw-caps. These self standing tubes are made of high-grade polypropylene, and include white printed graduations and marking area.
Important: Caps may be supplied loosely attached to allow effective sterilization. Product sterility is maintained by the sealed packaging. Once the sterile bag is opened, sterility of any unused tubes is not guaranteed.
Important: Caps may be supplied loosely attached to allow effective sterilization. Product sterility is maintained by the sealed packaging. Once the sterile bag is opened, sterility of any unused tubes is not guaranteed.
Features and Benefits
- Leak-proof, 1.25 turn screw-cap
- Meet 95kPa transportation guidelines
- Vivid, printed graduations
- Sterile, RNase, DNase, Pyrogen free
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio Transport/Mailing Tubes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.