Skip to Content

MTCD339038S

MTC Bio OptiWell Automation Reservoir

capacity 185 mL, 384 channel, polypropylene, sterile, case of 50 ea (10 bags of 5ea)

Synonym(s):

liquid reservoir, MTC Bio D3390-38S, liquid receptacle, reagent dish, reagent holder, reservoir

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41121522
Manufacturer/tradename:
MTC Bio D3390-38S
Capacity:
185 mL
Material:
clear , polypropylene
Sterility:
sterile
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

clear , polypropylene

sterility

sterile

feature

autoclavable, 384 channel

packaging

case of 50 ea (10 bags of 5ea)

manufacturer/tradename

MTC Bio D3390-38S

parameter

-80 °C temp. range

H

31.4 mm

capacity

185 mL

General description

OptiWell Reservoirs are compatible with a wide range of automated liquid handling equipment. Offered with 1 to 384 wells, they are useful in multiple applications, including mixing, fraction collection, plate replication, compound storage, and scale-up / scale-down methods. OptiWell Reservoirs conform with SLAS standard dimensions. They are made of chemically inert, low binding polypropylene to minimize excess residue/waste.

Features and Benefits

  • ANSI / SLAS 1-2004 Compliant
  • Certified RNase, DNase and pyrogen free
  • Robot friendly / standard dimensions
  • Autoclavable and freezable to -80°C
  • SLAS standard footprint

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library