MilliporeSigma
MTCM4025W
MTC™ Bio Inoculating Loops and Needles
flexible loop, capacity 10 μL, polypropylene, sterile, case of 1000 ea (individually wrapped)
Synonym(s):
culture needle, cutlure loop, MTC Bio M4025-W, sampling loop, sampling tool
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41161502
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
polypropylene
sterility
sterile
packaging
case of 1000 ea (individually wrapped)
manufacturer/tradename
MTC™ Bio M4025-W
capacity
10 μL
General description
Sterile disposable plastic loops and needles designed for use in quantitative procedures such as sampling, urine counts, and serial dilutions, as well as for bacterial inoculation. Because they are pre-sterilized and designed for single use, there is no risk of cross-contamination and no need for flaming. The unique hexagonal shape of the shaft makes it easier to grip, orientate, and guide through a smooth streaking process. The loop surfaces are extremely smooth with no rough edges and no flashing or burrs on the loop head. This results in the elimination of cutting or gouging of the agar surface.
Flexible Polypropylene Loops - Make it easier to maneuver them within Petri dishes, test tubes, or other confined spaces, improving the researcher′s ability to collect and transfer samples efficiently. The flexibility allows for more precise sampling from cultures, especially when working with small or delicate colonies that could be easily damaged by more rigid instruments.
Rigid Polystyrene Loops & Needles - Allow for more precise streaking patterns on agar plates, which is essential for isolating individual colonies or performing quantitative assays. They are also better suited for handling viscous or dense samples, where flexibility might impede the ability to effectively collect and transfer the sample.
Features and Benefits
  • Sterilized to SAL 10-6
  • Easy-grip hexagonal handle design
  • Flexible PP or Rigid PS available
  • Individually wrapped or peel packs of 20
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)