Skip to Content
MilliporeSigma

MTCP1001AS

MTC™ Bio Pure.Amp™ Aluminum Sealing Film

to fit standard PCR plates, sterile

Synonym(s):

MTC™ Bio Pure.Amp™ Aluminum Sealing Film, MTC Bio CP1001-AS, PCR plate seal, aluminimum membrane, plate sealing membrane, sealing membrane

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
MTC™ Bio P1001-A-S
Sterility:
sterile
Packaging:
pack of 100 ea (10 bags of 10 seals)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

sterile

packaging

pack of 100 ea (10 bags of 10 seals)

manufacturer/tradename

MTC™ Bio P1001-A-S

General description

PCR Sealing Film, Aluminum Foil, Adhesive, Pierceable, Sterile, 10 bags of 10 seals, 100/pk
Pure.Amp™ sealing films are compatible with all standard PCR plates and are guaranteed to keep evaporation rates below 5%.

Application

PCR

Features and Benefits

  • Certified RNase, DNase, DNA, & PCR inhibitor free

Legal Information

MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library