MilliporeSigma
MTCP1001ES
MTC™ Bio Pure.Amp™ ELISA Plate Sealing Film
transparent, sterile
Synonym(s):
MTC™ Bio Pure.Amp™ ELISA Plate Sealing Film, ELISA plate seal, MTC Bio C1001-E-S, low adhesive membrane, qPCR plate sealing membrance, sealing membrane
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
MTC™ Bio P1001-E-S
Material:
transparent
Sterility:
sterile
Packaging:
pack of 100 ea (10 bags of 10 seals)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
transparent
sterility
sterile
packaging
pack of 100 ea (10 bags of 10 seals)
manufacturer/tradename
MTC™ Bio P1001-E-S
General description
PureAmp Sealing Membranes compatible with all standard PCR plates for use in ELISA, EIA, and General Applications. Transparent film with low-adhesive EZ-peel, guaranteed to keep evaporation rates below 5%.
Sealing Film, Transparent, Easy Removal Low Adhesive, for ELISA, EIA, and General Applications, Sterile, 10 bags of 10 seals, 100/pk
Features and Benefits
  • Certified RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates <5%
  • Packed as 10 bags of 10 membranes
  • Temperature range -40° to 120°C
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)