MilliporeSigma
MTCP1001PCRS
MTC™ Bio Pure.Amp™ Transparent Sealing Film
to fit standard PCR plates, sterile
Synonym(s):
MTC™ Bio Pure.Amp™ Transparent Sealing Film, MTC Bio P1001-PCR-S, clear sealing membrane, plate seal, plate sealing film, sealing membrane
Select a Size
Change View
About This Item
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
MTC™ Bio P1001-PCR-S
Material:
transparent
Sterility:
sterile
Packaging:
pack of 100 ea (10 bags of 10 seals)
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
transparent
sterility
sterile
packaging
pack of 100 ea (10 bags of 10 seals)
manufacturer/tradename
MTC™ Bio P1001-PCR-S
General description
PCR Sealing Film, Transparent, Adhesive, Sterile, 10 bags of 10 seals, 100/pk
PureAmp™ standard adhesive transparent sealing films are compatible with all standard PCR plates and are guaranteed to keep evaporation rates below 5%.
Features and Benefits
  • RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates ≤ 5%
  • Inner packs of 10 membranes
  • -40° to 120°C
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)