MilliporeSigma
MTCP1001QS
MTC™ Bio Pure.Amp™ qPCR Sealing Film
to fit standard qPCR plates, standard adhesive, sterile
Synonym(s):
MTC™ Bio Pure.Amp™ qPCR Sealing Film, MTC Bio P1001-Q-S, adhesive membrane, plate seal, plate sealing film, sealing membrane
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122410
eCl@ss:
32120613
Manufacturer/tradename:
MTC™ Bio P1001-Q-S
Material:
clear (optically clear)
Sterility:
sterile
Packaging:
pack of 100 ea (10 bags of 10 seals)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear (optically clear)
sterility
sterile
packaging
pack of 100 ea (10 bags of 10 seals)
manufacturer/tradename
MTC™ Bio P1001-Q-S
General description
Pure.Amp™ Sealing Films are compatible with all standard PCR plates. Optically clear with standard adhesive, or non-tacky pressure activated adhesive, this plate sealing membrane is guaranteed to keep evaporation rates below 5%.
qPCR Sealing Film, Optically Clear, Standard Adhesive, Sterile, 10 bags of 10 seals, 100/pk
Features and Benefits
  • RNase, DNase, DNA, and PCR inhibitor free
  • Evaporation rates ≤ 5%
  • Inner packs of 10 membranes
  • -40° to 120°C
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)