MilliporeSigma
MTCP3010AY
MTC™ Bio Pure.Amp™™ PCR 8 Tube Strips
capacity 0.2 mL, flat cap (attached), yellow, pack of 120 ea
Synonym(s):
MTC™ Bio Pure.Amp™ PCR 8 Tube Strips, PCR sample storage tubes, sample storage tube, snap closure tube, MTC Bio P3010-A-Y
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121800
Manufacturer/tradename:
MTC™ Bio P3010-A-Y
Capacity:
0.2 mL
Material:
flat cap (attached), polypropylene , yellow
Packaging:
pack of 120 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
flat cap (attached), polypropylene , yellow
packaging
pack of 120 ea
manufacturer/tradename
MTC™ Bio P3010-A-Y
capacity
0.2 mL
General description
Pure.Amp™ 0.2mL PCR strips can be supplied with domed caps or flat caps and can be attached or supplied separately, color-coded tubes are easily identified at a glance, making them the ideal tube to track your samples. Strips are precision manufactured to maintain tight dimensional tolerances of ultra-thin wall thickness, for improved thermal transfer and are certified free of RNase, DNase, DNA and PCR inhibitors, and are made of heat-resistant medical-grade polypropylene.
Features and Benefits
  • RNase, DNase, DNA, and PCR inhibitor free
  • Ultra-thin, uniform wall thickness
Legal Information
MTC is a trademark of MTC Bio
Pure.Amp is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)