MilliporeSigma
MTCP4305R
MTC™ Bio Pipette Tips For ProPette™ LE Pipettor
maximum volume 5 mL, non-sterile, racked, pack of 144 ea (6 racks of 24ea)
Synonym(s):
MTC™ Bio Pipette Tips For ProPette™ LE Pipettor, pipetting tip, platic tip, MTC Bio P4305-R, dispensing tip
Select a Size
Change View
About This Item
UNSPSC Code:
41121600
eCl@ss:
32039090
Sterility:
non-sterile
Feature:
RNase and DNase free, graduations: no, racked
Packaging:
pack of 144 ea (6 racks of 24ea)
Manufacturer/tradename:
MTC™ Bio P4305-R
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear , polypropylene
sterility
non-sterile
feature
RNase and DNase free, graduations: no, racked
packaging
pack of 144 ea (6 racks of 24ea)
manufacturer/tradename
MTC™ Bio P4305-R
maximum volume
5 mL
General description
Non-filtered and filtered versions fit all 200µL pipettes that use standard tips. This includes Gilson, Eppendorf, and countless others. The non-filter tips will also fit many 100µL pipettes. Although they are “universal”, they are designed to fit MTC ™ and Propette™ LE pipettors.
Features and Benefits
  • RNase and DNase free
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)