Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41121514
eCl@ss:
32070201
Battery power:
lithium
Feature:
USB-C charging port
Manufacturer/tradename:
MTC™ Bio P6290
feature
USB-C charging port
packaging
pack of 1 ea
manufacturer/tradename
MTC™ Bio P6290
battery power
lithium
General description
Propette™ Digital Pipette Controllers add even more convenience and funcionality to the popular Propette™ Line. New buttons and an enhanced digital display allow users to set their max aspirate and dispense rates on the fly. Paired with Propette™ pressure-sensitive triggers, users can enjoy the most precise flow controls possible. The display also keeps constant track of the battery level, which can be charged via the convenient new USB-C port. The controller is lightweight, only 6.3oz/170g, and designed to fit comfortably in either the right or left hand. The natural feel of the ergonomic body design, the computer-balanced weight distribution, and the low-pressure fingertip control button all combine to provide a more comfortable pipetting experience. Includes the proprietary Quickstand retractable stand, which allows the instrument to be set down on the bench without risk of contamination, also USB-C charging Port and cable.
Features and Benefits
- Enhanced flow control - Max aspirate and dispense rates
- Independent aspiration and dispense speed settings
- Pressure sensitive triggers for even more control
- Improved QuickStand™ - Locks in place with a click
- USB-C charge port - No searching for specific chargers
Legal Information
MTC is a trademark of MTC Bio
ProPette is a trademark of MTC Bio
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.