MilliporeSigma
MTCR1020B
MTC™ Bio Storage Box With Hinged Lid
to hold, 100 x 1.5 mL tubes, blue, pack of 5 ea
Synonym(s):
MTC™ Bio Storage Box With Hinged Lid, tube rack, tube storage box, MTC Bio R1020-B
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122800
Manufacturer/tradename:
MTC™ Bio R1020-B
Feature:
to hold 100 x 1.5 mL tubes
Material:
blue , polypropylene
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
blue , polypropylene
feature
to hold 100 x 1.5 mL tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1020-B
General description
Polypropylene storage boxes suitable for refrigerator and freezer storage as low as -80˚C. 10×10 Format capable of holding up to 100 x 1.5 mL tubes, with an attached, translucent, hinged lid and numbered cavities.
Features and Benefits
  • Freezer storage as low as -80˚C
  • Translucent lids and numbered cavities
  • 10 × 10 format
  • Holds 100 x 1.5 mL tubes
  • Attached, hinged lid
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Choose from one of the most recent versions:
Certificates of Analysis (COA)