MilliporeSigma
MTCR1115
MTC™ Bio Test Tube Rack
to hold, 90 x 12-13 mm diameter FACS, or culture tubes, pack of 5 ea
Synonym(s):
tube holder, MTC Bio R1115, tube organizer, tube rack, tube storage
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
41122804
eCl@ss:
32032201
Manufacturer/tradename:
MTC™ Bio R1115
Feature:
autoclavable, to hold 90 x 12-13 mm diameter FACS, or culture tubes
Material:
blue , polypropylene
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
blue , polypropylene
feature
autoclavable, to hold 90 x 12-13 mm diameter FACS, or culture tubes
packaging
pack of 5 ea
manufacturer/tradename
MTC™ Bio R1115
General description
These autoclavable test tube racks are ideal for a variety of widely used laboratory tubes and vials. Manufactured in two sizes, these racks accommodate tubes ranging from 12-17mm in diameter, including test tubes, culture tubes, FACS tubes, large cryogenic vials, transport vials and more. Shipped in an economical flat pack, these racks snap together to create a sturdy and durable workstation. Molded alphanumeric grids along the sides assist in easy organization of samples. Available in 60 or 90 place, these heavy-duty polypropylene racks are autoclavable and can be safely used in water baths without floating.
Features and Benefits
  • 90-place (6 rows of 15) for 12-14mm tubes
  • 60-place (5 rows of 12) for 15-17mm tubes
  • Molded alphanumeric grids
  • Autoclavable polypropylene
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC™ Bio Test Tube Rack
Choose from one of the most recent versions:
Certificates of Analysis (COA)