Skip to Content
MilliporeSigma

MTCR1900

MTC™ Bio centrifuge tube rack

to hold, 6 x 175 / 225 or 500 mL centrifuge tubes, pack of 5 ea

Synonym(s):

tube holder, tube storage, MTC Bio R1900, tube organiser, tube rack

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41122804
Manufacturer/tradename:
MTC™ Bio R1900
Feature:
to hold 6 x 175 / 225 or 500 mL centrifuge tubes
Material:
blue , polypropylene
Packaging:
pack of 5 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

blue , polypropylene

feature

to hold 6 x 175 / 225 or 500 mL centrifuge tubes

packaging

pack of 5 ea

manufacturer/tradename

MTC™ Bio R1900

General description

Tube racks made of 100% polypropylene to hold 6 x 175 , 225 or 250 mL centrifuge tubes, can be snap-linked together to form as large as rack as is needed.

Features and Benefits

  • Ships flat
  • Holds 6 x 250mL, 175mL or 250mL tubes
  • Multiple racks can be linked together

Legal Information

MTC is a trademark of MTC Bio


Still not finding the right product?

Explore all of our products under MTC™ Bio centrifuge tube rack




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library